MilliporeSigma
  • Innate cytokine profiling of bovine alveolar macrophages reveals commonalities and divergence in the response to Mycobacterium bovis and Mycobacterium tuberculosis infection.
Innate cytokine profiling of bovine alveolar macrophages reveals commonalities and divergence in the response to Mycobacterium bovis and Mycobacterium tuberculosis infection.
Tuberculosis (Edinburgh, Scotland) (2014-06-03)
David A Magee, Kevin M Conlon, Nicolas C Nalpas, John A Browne, Chris Pirson, Claire Healy, Kirsten E McLoughlin, Jing Chen, H Martin Vordermeier, Eamonn Gormley, David E MacHugh, Stephen V Gordon
ABSTRACT
Despite sharing >99.9% genome sequence similarity at the nucleotide level, Mycobacterium tuberculosis and Mycobacterium bovis-the causative agents of human and bovine tuberculosis, respectively-exhibit distinct host preferences. M. bovis can cause disease in both cattle and humans yet rarely transmits between immuno-competent human hosts, while M. tuberculosis is a highly successful pathogen of humans that does not sustain in animal populations. Based on the key role played by alveolar macrophages during mycobacterial infection, we hypothesised that the immunological and pathological differences observed in cattle infected with virulent M. bovis and M. tuberculosis may have a basis in innate immune mechanisms; these differences, in turn, would be reflected at the macrophage mRNA and protein level. To investigate this, we have analysed the transcriptional profile of innate immune genes in bovine alveolar macrophages following 24 and 48 h infection with the genome-sequenced strains, M. bovis AF2122/97 and M. tuberculosis H37Rv. A bespoke multiplex ELISA was also used to quantify corresponding cytokine secretion in supernatants from the same infected alveolar macrophages. All cytokines showed similar significant patterns of expression (i.e., up- or down-regulation) at both the mRNA and protein levels in infected macrophages relative to parallel non-infected controls at the two time points (P ≤ 0.05). However, significant upregulation and downregulation of several innate immune genes-including TLR2, FOS, PIK3IP1, CCL4, IL1B, IL6 and TNF-and the CCL-4 protein was observed in the M. bovis-infected macrophages relative to the M. tuberculosis-infected macrophages 48 h post-infection (P ≤ 0.05). These results support the hypothesis that the divergent virulence of M. bovis and M. tuberculosis in cattle has a basis in innate immune mechanisms, which may contribute to host preference within the M. tuberculosis complex of strains.
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin, analytical standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)