MilliporeSigma
  • Therapeutic effects of fucoidan in 6-hydroxydopamine-lesioned rat model of Parkinson's disease: Role of NADPH oxidase-1.
Therapeutic effects of fucoidan in 6-hydroxydopamine-lesioned rat model of Parkinson's disease: Role of NADPH oxidase-1.
CNS neuroscience & therapeutics (2014-11-18)
Fei-Long Zhang, Yi He, Yan Zheng, Wen-Jing Zhang, Qi Wang, Yan-Jun Jia, Hai-Long Song, Hai-Ting An, Hao-Bo Zhang, Yan-Jing Qian, Yu-Long Tong, Lin Dong, Xiao-Min Wang
ABSTRACT
To explore the effect of fucoidan treatment on oxidative stress-mediated dopaminergic neuronal damage and its potential mechanisms. The effect of fucoidan was investigated in a 6-hydroxydopamine (6-OHDA) rat model of PD, an animal model considered appropriate for preclinical studies of PD therapy. The effects of fucoidan treatment on animal behavior and the survival ratio of dopaminergic neurons were investigated. We further observed the effect of fucoidan on microglia and the NADPH oxidases-1 (Nox1), a family of enzymes generating reactive oxygen species (ROS). We found that chronic fucoidan administration mitigated the motor dysfunction induced by 6-OHDA. Similarly, fucoidan reduced the loss of DA neurons in the SNc and DA fibers in the striatum in 6-OHDA-lesioned rats. Moreover, we found that fucoidan inhibited the 6-OHDA-stimulating expression of Nox1 in both tyrosine hydroxylase (TH)-positive neurons and non-TH-positive neurons, prevented Nox1-sensitive oxidative stress and cell damage in SNc neurons. Fucoidan also effectively inhibited nigral microglial activation. These results support the beneficial effect of fucoidan in 6-OHDA-lesioned rat model of PD. Fucoidan may suppress the Nox1-triggered oxidative stress in the SNc to protect DA neurons from 6-OHDA-induced toxicity and achieve its beneficial effect.
Product Description
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
DL-Tyrosine, 99%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-(−)-Fucose, ≥99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Supelco
2,4-Dinitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)