  • Search Results
  • Type, density, and presentation of grafted adhesion peptides on polysaccharide-based hydrogels control preosteoblast behavior and differentiation.
Type, density, and presentation of grafted adhesion peptides on polysaccharide-based hydrogels control preosteoblast behavior and differentiation.
Biomacromolecules (2015-01-30)
Jing Jing, Audrey Fournier, Anna Szarpak-Jankowska, Marc R Block, Rachel Auzély-Velty
ABSTRACT
In this work, cell-responsive polysaccharide hydrogels were prepared by a simple procedure based on the sequential bioconjugation and cross-linking of the polysaccharide backbone with bioactive peptides and poly(ethylene glycol)-bis(thiol) (PEG-(SH)2), respectively. Using thiol-ene reactions, we successfully functionalized hyaluronic acid (HA) and carboxymethylcellulose (CMC) with short and long peptides (5-mer and 15-mer derivatives, respectively) derived from adhesive proteins of bone extracellular matrix. The resulting HA-peptide and CMC-peptide conjugates with varying degrees of substitution were then carefully characterized by (1)H NMR spectroscopy to precisely control the peptide density into the hydrogels cross-linked with PEG-(SH)2. Preosteoblast seeded on the hydrogels with controlled identical stiffness spread in a manner that was strongly dependent on ligand density. Surprisingly, increasing the density of the adhesive peptide anchors did not result in a plateau of initial cell spreading but rather in a bell-shaped cell response that varies with the nature of both polysaccharide backbone and functional peptide. Placing the cells under optimal conditions for cell/hydrogel interaction, we showed that in HA hydrogels, the polysaccharide moiety is not solely a passive scaffold that presents the active peptides but is an active player in cell microenvironment to control and sustain cell activity.
Sigma-Aldrich
Deuterium oxide, 99 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, glass distilled
Sigma-Aldrich
Deuterium oxide, extra, 99.994 atom % D
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%