  • Search Results
  • Synthetic polyglutamylation of dual-functional MTX ligands for enhanced combined cytotoxicity of poly(I:C) nanoplexes.
Synthetic polyglutamylation of dual-functional MTX ligands for enhanced combined cytotoxicity of poly(I:C) nanoplexes.
Molecular pharmaceutics (2014-04-24)
Ulrich Lächelt, Valentin Wittmann, Katharina Müller, Daniel Edinger, Petra Kos, Miriam Höhn, Ernst Wagner
ABSTRACT
The antifolate drug methotrexate (MTX) can serve as a dual-functional ligand in antitumoral drug delivery, inducing both a folate receptor mediated cellular uptake and an intracellular cytotoxic action. Bioactivity of MTX however changes by conjugation; the activity can be affected by the hampered intracellular conversion to more potent poly-γ-glutamyl derivatives. Therefore, in a cancer combination therapy approach for the codelivery of cytotoxic dsRNA polyinosinic-polycytidylic acid poly(I:C), a set of molecularly precise oligo(ethanamino)amides were synthesized comprising poly(ethylene glycol) conjugated MTX ligands. The conjugates differed in the number of additional glutamic acid residues to investigate the effect of different degrees of synthetic "a priori" polyglutamylation. The bioactivity of these compounds concerning dihydrofolate reductase (DHFR) inhibition, cytotoxicity, nucleic acid binding potency, cellular uptake of poly(I:C) polyplexes, and combined antifolate/poly(I:C) toxicity was investigated. Synthetic polyglutamylation had a crucial impact on several stages of efficient poly(I:C) delivery and combined MTX cytotoxicity. DHFR inhibition of the conjugates significantly increased with increasing polyglutamate chain length. The library member with highest glutamylation degree even outperformed free MTX in direct comparison. Studies in KB cells showed the corresponding enhanced cytotoxicity by polyglutamylation. Also poly(I:C) polyplexes of the glutamylated MTX variants exhibited higher cellular uptake in the folate receptor positive cell line. Finally, a synergistic combined cytotoxicity of polyglutamylated MTX ligands and complexed poly(I:C) cargo was observed in transfected KB cells. The present structure-activity relationship study of MTX-based ligands pinpoints the concept of synthetic polyglutamylation as a promising approach for optimizing bioactivity of antifolate conjugates, which might be considered as a useful tool also in context of other drug delivery systems.
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Triisopropylsilane, 98%
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard