MilliporeSigma
CD8
Oncoimmunology (2015-05-08)
Benedikt Fritzsching, Joerg Fellenberg, Linda Moskovszky, Zoltan Sápi, Tibor Krenacs, Isidro Machado, Johannes Poeschl, Burkhard Lehner, Miklos Szendrõi, Antonio Llombart Bosch, Ludger Bernd, Monika Csóka, Gunhild Mechtersheimer, Volker Ewerbeck, Ralf Kinscherf, Pierre Kunz
ABSTRACT
Osteosarcoma is the most common primary bone tumor characterized by juvenile onset, tumor heterogeneity, and early pulmonary metastasis. Therapeutic improvement stagnates since more than two decades. Unlike major malignancies, biomarkers as prognostic factors at time of diagnosis are missing. Disease rareness hampers study recruitment of patient numbers sufficient to outweigh tumor heterogeneity. Here, we analyzed in a multicenter cohort the osteosarcoma microenvironment to reduce effects of tumor cell heterogeneity. We hypothesized that quantitative ratios of intratumoral CD8
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems