MilliporeSigma
  • TCAIM decreases T cell priming capacity of dendritic cells by inhibiting TLR-induced Ca2+ influx and IL-2 production.
TCAIM decreases T cell priming capacity of dendritic cells by inhibiting TLR-induced Ca2+ influx and IL-2 production.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-10)
Simone Z Vogel, Stephan Schlickeiser, Karsten Jürchott, Levent Akyuez, Julia Schumann, Christine Appelt, Katrin Vogt, Martina Schröder, Martin Vaeth, Friederike Berberich-Siebelt, Manfred B Lutz, Gerald Grütz, Birgit Sawitzki
ABSTRACT
We previously showed that the T cell activation inhibitor, mitochondrial (Tcaim) is highly expressed in grafts of tolerance-developing transplant recipients and that the encoded protein is localized within mitochondria. In this study, we show that CD11c(+) dendritic cells (DCs), as main producers of TCAIM, downregulate Tcaim expression after LPS stimulation or in vivo alloantigen challenge. LPS-stimulated TCAIM-overexpressing bone marrow-derived DC (BMDCs) have a reduced capacity to induce proliferation of and cytokine expression by cocultured allogeneic T cells; this is not due to diminished upregulation of MHC or costimulatory molecules. Transcriptional profiling also revealed normal LPS-mediated upregulation of the majority of genes involved in TLR signaling. However, TCAIM BMDCs did not induce Il2 mRNA expression upon LPS stimulation in comparison with Control-BMDCs. In addition, TCAIM overexpression abolished LPS-mediated Ca(2+) influx and mitochondrial reactive oxygen species formation. Addition of IL-2 to BMDC-T cell cocultures restored the priming capacity of TCAIM BMDCs for cocultured allogeneic CD8(+) T cells. Furthermore, BMDCs of IL-2-deficient mice showed similarly abolished LPS-induced T cell priming as TCAIM-overexpressing wild type BMDCs. Thus, TCAIM interferes with TLR4 signaling in BMDCs and subsequently impairs their T cell priming capacity, which supports its role for tolerance induction.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Supelco
Rotenone, PESTANAL®, analytical standard
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent