MilliporeSigma
  • A combination turbidity and supernatant microplate assay to rank-order the supersaturation limits of early drug candidates.
A combination turbidity and supernatant microplate assay to rank-order the supersaturation limits of early drug candidates.
Journal of pharmaceutical sciences (2014-07-30)
John S Morrison, Michelle J Nophsker, Roy J Haskell
ABSTRACT
A unique opportunity exists at the drug discovery stage to overcome inherently poor solubility by selecting drug candidates with superior supersaturation propensity. Existing supersaturation assays compare either precipitation-resistant or precipitation-inhibiting excipients, or higher-energy polymorphic forms, but not multiple compounds or multiple concentrations. Furthermore, these assays lack sufficient throughput and compound conservation necessary for implementation in the discovery environment. A microplate-based combination turbidity and supernatant concentration assay was therefore developed to determine the extent to which different compounds remain in solution as a function of applied concentration in biorelevant media over a specific period of time. Dimethyl sulfoxide stock solutions at multiple concentrations of four poorly soluble, weak base compounds (Dipyridamole, Ketoconazole, Albendazole, and Cinnarizine) were diluted with pH 6.5 buffer as well as FaSSIF. All samples were monitored for precipitation by turbidity at 600 nm over 1 h and the final supernatant concentrations were measured. The maximum supersaturation ratio was calculated from the supersaturation limit and the equilibrium solubility in each media. Compounds were rank-ordered by supersaturation ratio: Ketoconazole > Dipyridamole > Cinnarizine ∼ Albendazole. These in vitro results correlated well with oral AUC ratios from published in vivo pH effect studies, thereby confirming the validity of this approach.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Albendazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
USP
Albendazole, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Dipyridamole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets