  • Search Results
  • Mast cells control the expansion and differentiation of IL-10-competent B cells.
Mast cells control the expansion and differentiation of IL-10-competent B cells.
Journal of immunology (Baltimore, Md. : 1950) (2014-10-01)
Francesca Mion, Federica D'Incà, Luca Danelli, Barbara Toffoletto, Carla Guarnotta, Barbara Frossi, Alessia Burocchi, Alice Rigoni, Norbert Gerdes, Esther Lutgens, Claudio Tripodo, Mario P Colombo, Juan Rivera, Gaetano Vitale, Carlo E Pucillo
ABSTRACT
The discovery of B cell subsets with regulatory properties, dependent on IL-10 production, has expanded our view on the mechanisms that control inflammation. Regulatory B cells acquire the ability to produce IL-10 in a stepwise process: first, they become IL-10 competent, a poised state in which B cells are sensitive to trigger signals but do not actually express the Il-10 gene; then, when exposed to appropriate stimuli, they start producing IL-10. Even if the existence of IL-10-competent B cells is now well established, it is not yet known how different immune cell types cross talk with B cells and affect IL-10-competent B cell differentiation and expansion. Mast cells (MCs) contribute to the differentiation and influence the effector functions of various immune cells, including B lymphocytes. In this study, we explored whether MCs could play a role in the expansion of IL-10-competent B cells and addressed the in vivo relevance of MC deficiency on the generation of these cells. We show that MCs can expand IL-10-competent B cells, but they do not directly induce IL-10 production; moreover, the absence of MCs negatively affects IL-10-competent B cell differentiation. Noteworthy, our findings reveal that the CD40L/CD40 axis plays a significant role in MC-driven expansion of IL-10-competent B cells in vitro and highlight the importance of MC CD40L signaling in the colon.
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture