  • Search Results
  • RNASET2 tag SNP but not CCR6 polymorphisms is associated with autoimmune thyroid diseases in the Chinese Han population.
RNASET2 tag SNP but not CCR6 polymorphisms is associated with autoimmune thyroid diseases in the Chinese Han population.
BMC medical genetics (2015-05-01)
Xiao-jun Chen, Xiao-hua Gong, Ni Yan, Shuai Meng, Qiu Qin, Yan-Fei Jiang, Hai-Yan Zheng, Jin-an Zhang
ABSTRACT
Polymorphisms of the CC chemokine receptor 6 (CCR6) and RNASET2 tag SNP have been shown to be associated with the susceptibility to several immune-related diseases. This study was conducted to identify the association of CCR6 and RNASET2 tag SNP with autoimmune thyroid diseases (AITDs) in the Chinese Han population. We enrolled 1061 patients with AITDs, including 701 patients with Graves' disease (GD) and 360 patients with Hashimoto's thyroiditis (HT), and 938 healthy individuals for a case-control genetic association study. Three CCR6 single nucleotides polymorphisms (SNPs) (rs3093023/rs3093024/rs6902119) and one tagging SNP (rs9355610) within RNASET2 gene were selected for genotyping by multiplex polymerase chain reaction (PCR) and ligase detection reaction (LDR). The frequency of rs9355610 genotypes in the patients with GD differed significantly from that in the controls (p = 0.017). The frequency of the minor G allele of rs9355610 was significantly higher in the GD patients than in the healthy controls (p = 0.005, OR = 1.225, 95% CI:1.063-1.412). However, we could not find significant differences in the genotype or allele frequencies of HT patients compared with healthy controls. After gender stratification, the frequency of the minor G allele in both male and female GD patients was significantly higher than that in the healthy controls (p = 0.036, OR = 1.308, 95% CI:1.017-1.684 ; p = 0.048, OR = 1.19, 95% CI:1.001-1.413; respectively);. Furthermore, the frequency of haplotype AT in GD patients was significantly lower than that in their control groups (p = 0.003) and showed a protective effect against GD (OR = 0.806, 95% CI: 0.699-0.929). The frequency of haplotype GT in GD patients was significantly higher than that in their control groups (p = 0.048), indicating that GT was the risk haplotype to GD (OR = 1.267, 95% CI: 1.001-1.603). There were no significant differences in the allele or genotype frequencies of three SNPs of CCR6 (rs3093023/rs3093024/ rs6902119) gene between GD patients, HT patients and controls. Our results suggest that the rs9355610 tag SNP of RNASET2 gene is positively associated with susceptibility to GD in the Chinese Han population. No association was found for the tested CCR6 SNPs.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered