  • Search Results
  • Evolution of watermelon fruit physicochemical and phytochemical composition during ripening as affected by grafting.
Evolution of watermelon fruit physicochemical and phytochemical composition during ripening as affected by grafting.
Food chemistry (2014-07-21)
G A Soteriou, M C Kyriacou, A S Siomos, D Gerasopoulos
ABSTRACT
Flesh reflectance colorimetry, mechanical texture analysis, pH, titratable acidity (TA), and soluble solid (SS), soluble carbohydrate, lycopene and citrulline content of watermelon fruit were assessed throughout ripening (30-50 days post-anthesis; dpa) in grafted and self-rooted plants. Grafting increased firmness, TA, and lycopene content though it delayed its peak. Lycopene content was mostly ripening-dependant, highly correlated and synchronous with changes in pulp chroma (C) and colour a. The sweetness was affected only by ripening. However, total sugars and SS peaked later in fruit of grafted plants than in non-grafted ones, and significant interaction of ripening with grafting was observed. Citrulline content increased with ripening in fruit of grafted plants, reaching a peak at 45 dpa; whereas in non-grafted ones it was unchanged between 30 and 45 dpa and declined at 50 dpa. As ripening overall was retarded by grafting, fruit quality of grafted watermelon may benefit from belated harvest.
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O