  • Search Results
  • 17-DMAG induces Hsp70 and protects the auditory hair cells from kanamycin ototoxicity in vitro.
17-DMAG induces Hsp70 and protects the auditory hair cells from kanamycin ototoxicity in vitro.
Neuroscience letters (2015-01-06)
Yun Liu, Yang Yu, Hanqi Chu, Dan Bing, Shaoli Wang, Liangqiang Zhou, Jin Chen, Qingguo Chen, Chunchen Pan, Yanbo Sun, Yonghua Cui
ABSTRACT
Heat shock protein 70 (Hsp70) has been known to be able to play a protective role in the cochlea. The aim of this study was to investigate whether geldanamycin hydrosoluble derivative 17-(dimethylaminoethylamino)-17-demethoxygeldanamycin (17-DMAG) has the ability to induce Hsp70 up-regulation to protect hair cells from kanamycin-induced ototoxicity in vitro. The organ of Corti (OC) explants were isolated from mice at postnatal day 3-5. Then, the explants were exposed to kanamycin with or without pre-incubation with 17-DMAG. The expression of Hsp70 was assessed by reverse transcription-quantitative polymerase chain reaction, ELISA, and immunofluorescent staining. The surviving hair cells were examined by phalloidin labeling and were counted. We found that Hsp70 expression in the explants after pre-incubation with 17-DMAG was significantly increased at both mRNA and protein levels. Immunofluorescent staining showed that Hsp70 was mainly located in the auditory hair cells. Compared with kanamycin group, the loss of hair cells was inhibited significantly in 17-DMAG+kanamycin group. Our study demonstrated that 17-DMAG induces Hsp70 in the hair cells, and has a significant protective effect against kanamycin ototoxicity in vitro. 17-DMAG has the possibility to be a safe and effective anti-ototoxic drug.
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, peptide Amanita phalloides
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Kanamycin sulfate Streptomyces kanamyceticus, powder, γ-irradiated