  • Search Results
  • Hybrid paclitaxel and gold nanorod-loaded human serum albumin nanoparticles for simultaneous chemotherapeutic and photothermal therapy on 4T1 breast cancer cells.
Hybrid paclitaxel and gold nanorod-loaded human serum albumin nanoparticles for simultaneous chemotherapeutic and photothermal therapy on 4T1 breast cancer cells.
ACS applied materials & interfaces (2015-03-15)
Donna V Peralta, Zahra Heidari, Srikanta Dash, Matthew A Tarr
ABSTRACT
The use of human serum albumin nanoparticles (HSAPs) as a drug carrier system for cancer treatment has proven successful through current marketable clinical formulations. Despite this success, there is a current lack of multifunctional HSAPs, which offer combinational therapies of more than one proven technique. Gold nanorods (AuNRs) have also shown medicinal promise due to their photothermal therapy capabilities. In this study, a desolvation and cross-linking approach was employed to successfully encapsulate gold nanorods into HSAPs simultaneously with the chemotherapeutic drug paclitaxel (PAC); forming PAC-AuNR-HSAPs with desirable overall particle sizes of 299 ± 6 nm. The loading efficiency of paclitaxel into PAC-AuNR-HSAPs reached up to 3 μg PAC/mg HSA. The PAC-AuNR-HSAPs experienced photothermal heating; with the bulk particle solution reaching up to 46 °C after 15 min of near-IR laser exposure. This heat increase marked the successful attainment of the temperature necessary to cause severe cellular hyperthermia and necrosis. The encasement strategy facilitated a colloidal hybrid treatment system capable of enhanced permeability and retention effects, photothermal ablation of cancer cells, and release of the active paclitaxel of up to 188 ng (from PAC-AuNR-HSAPs created with 30 mg HSA) in a single 15 min irradiation session. When treated with PAC-AuNR-HSAPs containing 20 μg PAC/mL particle solution, 4T1 mouse breast cancer cells experienced ∼82% cell death without irradiation and ∼94% cell death after just one irradiation session. The results for PAC-AuNR-HSAPs were better than that of free PAC, which only killed ∼77% of the cells without irradiation and ∼80% with irradiation. The hybrid particle system also lends itself to future customizable external functionalities via conjugated targeting ligands, such as antibodies. Internal entrapment of patient tailored medication combinations are also possible with this combination treatment platform, which may result in improved quality of life for those undergoing treatment.
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%