MilliporeSigma
  • The uric acid transporter SLC2A9 is a direct target gene of the tumor suppressor p53 contributing to antioxidant defense.
The uric acid transporter SLC2A9 is a direct target gene of the tumor suppressor p53 contributing to antioxidant defense.
Oncogene (2014-05-27)
Y Itahana, R Han, S Barbier, Z Lei, S Rozen, K Itahana
ABSTRACT
Only humans and higher primates have high uric acid blood levels. Although high uric acid causes gout, it has been linked with human longevity because of its hypothetical antioxidant function. Recent studies reveal that p53 has significant roles in cellular metabolism. One example of this is an antioxidant function that potentially contributes to tumor suppression. Here, we reported a first beneficial link between p53 and uric acid. We identified the uric acid transporter SLC2A9 (also known as GLUT9) as a direct p53 target gene and a key downstream effector in the reduction of reactive oxygen species (ROS) through transporting uric acid as a source of antioxidant. Oxidative stress induced SLC2A9 expression in a p53-dependent manner, and inhibition of SLC2A9 by small interfering RNA (siRNA) or anti-gout drugs such as probenecid significantly increased ROS levels in an uric acid-dependent manner and greatly sensitized cancer cells to chemotherapeutic drugs. Conversely, expression of SLC2A9 reduced ROS and protected against DNA damage and cell death, suggesting its antioxidant function. The increased production of ROS because of p53 loss was rescued by SLC2A9 expression. Furthermore, decreased SLC2A9 expression was observed in several cancer types and was associated with a poorer prognosis. Our findings suggest that the p53-SLC2A9 pathway is a novel antioxidant mechanism that uses uric acid to maintain ROS homeostasis and prevent accumulation of ROS-associated damage that potentially contributes to cancer development.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzbromarone, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Benzbromarone, analytical standard
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Hydroxytamoxifen, analytical standard, (E) and (Z) isomers (50:50)
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
S-Nitrosoglutathione, ≥97%
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Diamide
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
4-Hydroxytamoxifen, ≥70% Z isomer (remainder primarily E-isomer)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%