  • Search Results
  • Krüppel-like factor 6 interferes with cellular transformation induced by the H-ras oncogene.
Krüppel-like factor 6 interferes with cellular transformation induced by the H-ras oncogene.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-09-13)
Lucas Daniel Trucco, Verónica Andreoli, Nicolás Gonzalo Núñez, Mariana Maccioni, José Luis Bocco
ABSTRACT
KLF6 is a member of the Krüppel-like factor family of transcription factors, with diverse roles in the regulation of cell physiology, including proliferation, signal transduction, and apoptosis. Mutations or down-regulation of KLF6 have been described in several human cancers. In this work, we found that KLF6-knockdown resulted in the formation of transformed foci and allowed the spontaneous conversion of NIH3T3 cells to a tumorigenic state. We further assessed the role of KLF6 in the context of oncogenic Ras. We showed that KLF6 was up-regulated by H-Ras(G12V) expression in a Jun N-terminal kinase (JNK)-dependent manner, correlated with enhanced klf6 promoter activity. We found that ectopic KLF6 expression induced a G1-phase cell cycle arrest, thereby decreasing the cell proliferation rate. In addition, constitutive KLF6 expression impaired H-Ras(G12V)-mediated loss of density-dependent growth inhibition and anchorage-independent growth. Moreover, growth of H-Ras(G12V)-driven tumors was reduced in mice challenged with cells stably expressing KLF6. KLF6 expression correlated with the up-regulation of p21, whereas neither p53 induction nor apoptotic cell death was detected. Further, p21 knockdown impaired KLF6-induced cell cycle arrest. These findings provide novel evidence highlighting KLF6 function in response to malignant transformation, suggesting the relevance of KLF6 in controlling cell proliferation and hindering tumorigenesis.
Product Description
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
SAFC
Methotrexate
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Methotrexate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Phenazine methosulfate, ≥90% (UV)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O