MilliporeSigma
  • Adipose hypothermia in obesity and its association with period homolog 1, insulin sensitivity, and inflammation in fat.
Adipose hypothermia in obesity and its association with period homolog 1, insulin sensitivity, and inflammation in fat.
PloS one (2014-11-15)
Masaya Yamaoka, Norikazu Maeda, Yasunori Takayama, Ryohei Sekimoto, Yu Tsushima, Keisuke Matsuda, Takuya Mori, Kana Inoue, Hitoshi Nishizawa, Makoto Tominaga, Tohru Funahashi, Iichiro Shimomura
ABSTRACT
Visceral fat adiposity plays an important role in the development of metabolic syndrome. We reported previously the impact of human visceral fat adiposity on gene expression profile of peripheral blood cells. Genes related to circadian rhythm were highly associated with visceral fat area and period homolog 1 (PER1) showed the most significant negative correlation with visceral fat area. However, regulation of adipose Per1 remains poorly understood. The present study was designed to understand the regulation of Per1 in adipose tissues. Adipose Per1 mRNA levels of ob/ob mice were markedly low at 25 and 35 weeks of age. The levels of other core clock genes of white adipose tissues were also low in ob/ob mice at 25 and 35 weeks of age. Per1 mRNA was mainly expressed in the mature adipocyte fraction (MAF) and it was significantly low in MAF of ob/ob mice. To examine the possible mechanisms, 3T3-L1 adipocytes were treated with H2O2, tumor necrosis factor-α (TNF-α), S100A8, and lipopolysaccharide (LPS). However, no significant changes in Per1 mRNA level were observed by these agents. Exposure of cultured 3T3-L1 adipocytes to low temperature (33°C) decreased Per1 and catalase, and increased monocyte chemoattractant protein-1 (Mcp-1) mRNA levels. Hypothermia also worsened insulin-mediated Akt phosphorylation in 3T3-L1 adipocytes. Finally, telemetric analysis showed low temperature of adipose tissues in ob/ob mice. In obesity, adipose hypothermia seems to accelerate adipocyte dysfunction.
Product Description
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
7-Amino-1,3-naphthalenedisulfonic acid monopotassium salt monohydrate, BioReagent, suitable for fluorescence, ≥98.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Adenosine, European Pharmacopoeia (EP) Reference Standard