  • Search Results
  • Comparison of sample preparation methods for the quantitative analysis of eicosanoids and other oxylipins in plasma by means of LC-MS/MS.
Comparison of sample preparation methods for the quantitative analysis of eicosanoids and other oxylipins in plasma by means of LC-MS/MS.
Analytical and bioanalytical chemistry (2014-12-30)
Annika I Ostermann, Ina Willenberg, Nils Helge Schebb
ABSTRACT
Oxylipins are potent lipid mediators. For the evaluation of their biological roles, several LC-MS based methods have been developed. While these methods are similar, the described sample preparation procedures for the extraction of oxylipins differ considerably. In order to deduce the most appropriate method for the analysis of non-esterified oxylipins in human plasma, we evaluated the performance of seven established sample preparation procedures. Six commonly used solid phase extraction (SPE) and one liquid-liquid extraction (LLE) protocol were compared based on the recovery of 13 added internal standards, extraction efficacy of oxylipins from plasma and reduction of ion-suppressing matrix. Dramatic differences in the performance in all three parameters were found. LLE with ethyl acetate was overall not a sufficient sample preparation strategy. The protocols using Oasis- and StrataX-material insufficiently removed interfering matrix compounds. Extraction efficacy of oxylipins on anion-exchanging BondElut cartridges was low, while removal of matrix was nearly perfect. None of the protocols led to a high extraction efficacy of analytes while removing all interfering matrix components. However, SPE on a C18-material with removal of matrix by water and n-hexane prior elution with methyl formate showed the best performance for the analysis of a broad spectrum of oxylipins in plasma.
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
SAFC
Acetic acid, glacial
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG