  • Search Results
  • Lipopeptides are signaled by Toll-like receptor 1, 2 and 6 in endolysosomes.
Lipopeptides are signaled by Toll-like receptor 1, 2 and 6 in endolysosomes.
International immunology (2014-05-27)
Yuji Motoi, Takuma Shibata, Koichiro Takahashi, Atsuo Kanno, Yusuke Murakami, Xiaobing Li, Tadashi Kasahara, Kensuke Miyake
ABSTRACT
Toll-like receptors (TLRs) recognize a variety of microbial products and induce defense responses. Pathogen sensing by TLRs occurs either on the cell surface or in endolysosomes. TLR-dependent responses are greatly influenced by the site of pathogen sensing. TLR heterodimers TLR1/TLR2 and TLR2/TLR6 recognize tri- or diacylated microbial lipopeptides, respectively. Although TLR1, 2 and 6 are believed to localize on the cell surface of immune cells, little is known about where lipopeptides are signaled. In this study, we established mAbs to TLR1, 2 and 6. TLR1, 2 and 6 were expressed on the surface of B cells, monocytes and dendritic cells in a manner dependent on a TLR-specific chaperone PRAT4A (protein associated with TLR4 A). Cell surface localization of TLR1 or TLR6 was not necessarily required for TLR2 response. Furthermore, a dynamin inhibitor 'Dynasore' abolished the lipopeptide responses by preventing lipopeptide internalization into LAMP-1 and LAMP-2 positive compartments. Our findings suggest that lipopeptides elicit TLR1/2 and TLR2/6 signaling in the endolysosomes, but not on the cell surface.
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dynasore hydrate
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)