  • Search Results
  • The BCL9-2 proto-oncogene governs estrogen receptor alpha expression in breast tumorigenesis.
The BCL9-2 proto-oncogene governs estrogen receptor alpha expression in breast tumorigenesis.
Oncotarget (2014-08-26)
Nathalie Zatula, Maria Wiese, Jens Bunzendahl, Walter Birchmeier, Christina Perske, Annalen Bleckmann, Felix H Brembeck
ABSTRACT
The majority of human breast cancers express estrogen receptor alpha (ER), which is important for therapy with anti-estrogens. Here we describe the role of BCL9-2, a proto-oncogene previously characterized as co-activator of Wnt/ß-catenin signaling, for mammary tumorigenesis in mice and human. ER positive human breast cancers showed overexpression of BCL9-2 and tamoxifen treated patients with high BCL9-2 demonstrated a better survival. BCL9-2 was upregulated during puberty and pregnancy in normal mammary epithelia, but downregulated in the involuted gland. BCL9-2 overexpression in vivo delayed the mammary involution and induced alveolar hyperplasia. Moreover, aged BCL9-2 transgenic mice developed ductal-like mammary tumors with high nuclear ER expression. We found, that primary cell cultures of BCL9-2 breast tumors responded to tamoxifen treatment. Moreover, BCL9-2 regulated the expression of ER and the proliferation of human breast cancer cells independently of ß-catenin. Finally, we describe a novel mechanism, how BCL9-2 regulates ER transcription by interaction with Sp1 through the proximal ESR1 gene promoter. In summary, BCL9-2 induces ER positive breast cancers in vivo, regulates ER expression by a novel ß-catenin independent mechanism in breast cancer cells, and might predict the therapy response to tamoxifen treatment.
Product Description
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Dexamethasone, VETRANAL®, analytical standard
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.