  • Search Results
  • Transformation of the intestinal epithelium by the MSI2 RNA-binding protein.
Transformation of the intestinal epithelium by the MSI2 RNA-binding protein.
Nature communications (2015-03-17)
Shan Wang, Ning Li, Maryam Yousefi, Angela Nakauka-Ddamba, Fan Li, Kimberly Parada, Shilpa Rao, Gerard Minuesa, Yarden Katz, Brian D Gregory, Michael G Kharas, Zhengquan Yu, Christopher J Lengner
ABSTRACT
The MSI2 RNA-binding protein is a potent oncogene playing key roles in haematopoietic stem cell homeostasis and malignant haematopoiesis. Here we demonstrate that MSI2 is expressed in the intestinal stem cell compartment, that its expression is elevated in colorectal adenocarcinomas, and that MSI2 loss-of-function abrogates colorectal cancer cell growth. MSI2 gain-of-function in the intestinal epithelium in a drug-inducible mouse model is sufficient to phenocopy many of the morphological and molecular consequences of acute loss of the APC tumour suppressor in the intestinal epithelium in a Wnt-independent manner. Transcriptome-wide RNA-binding analysis indicates that MSI2 acts as a pleiotropic inhibitor of known intestinal tumour suppressors including Lrig1, Bmpr1a, Cdkn1a and Pten. Finally, we demonstrate that inhibition of the PDK-AKT-mTORC1 axis rescues oncogenic consequences of MSI2 induction. Taken together, our findings identify MSI2 as a central component in an unappreciated oncogenic pathway promoting intestinal transformation.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Tamoxifen, analytical standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Supelco
Doxycycline hyclate, VETRANAL®, analytical standard
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Doxycycline hyclate
Doxycycline hyclate, European Pharmacopoeia (EP) Reference Standard
USP
Doxycycline hyclate, United States Pharmacopeia (USP) Reference Standard
Supelco
Doxycycline hyclate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
DL-Dithiothreitol solution, 1 M in H2O