  • Search Results
  • Exploring the impact of drug properties on the extent of intestinal lymphatic transport - in vitro and in vivo studies.
Exploring the impact of drug properties on the extent of intestinal lymphatic transport - in vitro and in vivo studies.
Pharmaceutical research (2014-11-28)
Emma Lawless, Brendan T Griffin, Aoife O'Mahony, Caitriona M O'Driscoll
ABSTRACT
Intestinal lymphatic transport of specific lipophilic drugs offers therapeutic advantages and maximises oral bioavailability. The aims of this study were; to compare intestinal lymphatic transport of a range of drugs and to investigate the influence of cyclosporine A on the mechanism/extent of lymphatic transport. Caco2 cells and an anaesthetised mesenteric lymphatic cannulated rat model were used for in vitro and in vivo studies. Lymphatic transport of three lipophilic drugs was directly compared in a long chain fatty acid formulation. In addition, the impact of cyclosporine A on triglyceride turnover was evaluated in vivo and in vitro. The extent of intestinal lymphatic transport in rats was positively correlated with drug solubility in triglyceride and negatively correlated with drug aqueous solubility. Cyclosporine A displayed non-linear lymphatic transport kinetics and reduced intestinal lymph triglyceride. In vitro experiments indicated that the cellular processes affected were intracellular lipid processing and/or lipid secretion. The linear correlations obtained using a range of lipophilic drugs confirm that the simplified approach of determining aqueous or triglyceride drug solubility is useful in predicting the extent of lymphatic transport. In vitro experiments correlated with in vivo observations, demonstrating the usefulness of the Caco-2 model for mechanistic investigations.
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Sodium hydroxide, beads, 20-40 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid