  • Search Results
  • Examination of the early infection stages of koi herpesvirus (KHV) in experimentally infected carp, Cyprinus carpio L. using in situ hybridization.
Examination of the early infection stages of koi herpesvirus (KHV) in experimentally infected carp, Cyprinus carpio L. using in situ hybridization.
Journal of fish diseases (2014-06-14)
S J Monaghan, K D Thompson, A Adams, J Kempter, S M Bergmann
ABSTRACT
Koi herpesvirus (KHV) causes a highly infectious disease afflicting common carp and koi, Cyprinus carpio L. Various molecular and antibody-based detection methods have been used to elucidate the rapid attachment and dissemination of the virus throughout carp tissues, facilitating ongoing development of effective diagnostic approaches. In situ hybridization (ISH) was used here to determine the target tissues of KHV during very early infection, after infecting carp with a highly virulent KHV isolate. Analysis of paraffin-embedded tissues (i.e. gills, skin, spleen, kidney, gut, liver and brain) during the first 8 h and following 10 days post-infection (hpi; dpi) revealed positive signals in skin mucus, gills and gut sections after only 1 hpi. Respiratory epithelial cells were positive as early as 2 hpi. Viral DNA was also detected within blood vessels of various tissues early in the infection. Notable increases in signal abundance were observed in the gills and kidney between 5 and 10 dpi, and viral DNA was detected in all tissues except brain. This study suggests that the gills and gut play an important role in the early pathogenesis of this Alloherpesvirus, in addition to skin, and demonstrates ISH as a useful diagnostic tool for confirmation of acutely infected carp.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Digoxigenin, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Supelco
Digoxigenin, analytical standard