  • Search Results
  • In situ evaluation of oxidative stress and immunological parameters as ecotoxicological biomarkers in a novel sentinel species (Mimachlamys varia).
In situ evaluation of oxidative stress and immunological parameters as ecotoxicological biomarkers in a novel sentinel species (Mimachlamys varia).
Aquatic toxicology (Amsterdam, Netherlands) (2015-02-24)
Thomas Milinkovitch, Paco Bustamante, Valérie Huet, Anaïs Reigner, Carine Churlaud, Hélène Thomas-Guyon
ABSTRACT
Although the variegated scallop Mimachlamys varia seems to be a suitable sentinel species for contaminant monitoring, no study has identified biomarkers in this species. In order to fill this gap, this study conducted an in situ biomarker approach. M. varia were collected in contaminated and uncontaminated areas and responsiveness of oxidative stress and immunological biomarkers was evaluated in the digestive gland. In parallel, 14 trace element concentrations were evaluated in the same organ. Superoxide dismutase activity and malondialdehyde content responded efficiently to in situ contamination when a certain degree of contamination was reached. Laccase-type phenoloxidase showed a high sensitivity but saturation of the response was highlighted for the highest contaminations. Additionally, correlations were found between biomarkers and trace element concentrations. Taken together, results showed that biomarker approach conducted in the digestive gland of M. varia represents a sensitive analytical tool to highlight ecotoxicological issues in coastal marine ecosystems.
Sigma-Aldrich
p-Phenylenediamine, 98% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
p-Phenylenediamine, ≥99.0% (GC/NT)
Sigma-Aldrich
p-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
SAFC
Acetic acid, glacial
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%