  • Search Results
  • Extracellular matrix modulates the biological effects of melatonin in mesenchymal stem cells.
Extracellular matrix modulates the biological effects of melatonin in mesenchymal stem cells.
The Journal of endocrinology (2014-09-12)
Fan He, Xiaozhen Liu, Ke Xiong, Sijin Chen, Long Zhou, Wenguo Cui, Guoqing Pan, Zong-Ping Luo, Ming Pei, Yihong Gong
ABSTRACT
Both self-renewal and lineage-specific differentiation of mesenchymal stem cells (MSCs) are triggered by their in vivo microenvironment including the extracellular matrix (ECM) and secreted hormones. The ECM may modulate the physiological functions of hormones by providing binding sites and by regulating downstream signaling pathways. Thus, the purpose of this study was to evaluate the degree of adsorption of melatonin to a natural cell-deposited ECM and the effects of this interaction on the biological functions of melatonin in human bone marrow-derived MSCs (BM-MSCs). The fibrillar microstructure, matrix composition, and melatonin-binding affinity of decellularized ECM were characterized. The cell-deposited ECM improved melatonin-mediated cell proliferation by 31.4%, attenuated accumulation of intracellular reactive oxygen species accumulation, and increased superoxide dismutase (SOD) mRNA and protein expression. Interaction with ECM significantly enhanced the osteogenic effects of melatonin on BM-MSCs by increasing calcium deposition by 30.5%, up-regulating osteoblast-specific gene expression and down-regulating matrix metalloproteinase (MMP) expression. The underlying mechanisms of these changes in expression may involve intracellular antioxidant enzymes, because osteoblast-specific genes were down-regulated, whereas MMP expression was up-regulated, in the presence of SOD-specific inhibitors. Collectively, our findings indicate the importance of native ECM in modulating the osteoinductive and antioxidant effects of melatonin and provide a novel platform for studying the biological actions of growth factors or hormones in a physiologically relevant microenvironment. Moreover, a better understanding of the enhancement of MSC growth and osteogenic differentiation resulting from the combination of ECM and melatonin could improve the design of graft substitutes for skeletal tissue engineering.
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Manganese, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 98.7%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Manganese, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.008mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Manganese, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%