  • Search Results
  • Induction of complementary function reductase enzymes in colon cancer cells by dithiole-3-thione versus sodium selenite.
Induction of complementary function reductase enzymes in colon cancer cells by dithiole-3-thione versus sodium selenite.
Journal of biochemical and molecular toxicology (2014-09-03)
Melanie M Erzinger, Cédric Bovet, Anuli Uzozie, Shana J Sturla
ABSTRACT
Cellular induction of reductase enzymes can alter the susceptibility of cells toward drugs and chemicals. In this study, we compared the capacity of a single dose of sodium selenite and 3H-1,2-dithiole-3-thione (D3T) to influence the drug-relevant reducing capacity of HT29 cells over time, and defined the protein-specific contribution to this activity on the basis of selected reaction monitoring mass spectrometry. Thioredoxin reductase 1 (TrxR1) protein levels and activity were inducible up to 2.2-fold by selenium. In contrast, selenium had only a minor influence on prostaglandin reductase 1 (PTGR1) and quinone oxidoreductase 1 (NQO1) activity and protein levels. D3T, a strong Nrf2 inducer, induced all the reductases and additionally increased the cytotoxicity of hydroxymethylacylfulvene, a bioreductive DNA-alkylating drug. The data and experimental approaches allow one to define induction potency for reductase enzymes PTGR1, TrxR1, and NQO1 in HT29 cells and link these to changes in drug cytotoxicity.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%