  • Search Results
  • DNA amplification in neutral liposomes for safe and efficient gene delivery.
DNA amplification in neutral liposomes for safe and efficient gene delivery.
ACS nano (2014-04-24)
Sangmin Lee, Heebeom Koo, Jin Hee Na, Kyung Eun Lee, Seo Young Jeong, Kuiwon Choi, Sun Hwa Kim, Ick Chan Kwon, Kwangmeyung Kim
ABSTRACT
In general, traditional gene carriers contain strong cationic charges to efficiently load anionic genes, but this cationic character also leads to destabilization of plasma membranes and causes severe cytotoxicity. Here, we developed a PCR-based nanofactory as a safe gene delivery system. A few template plasmid DNA can be amplified by PCR inside liposomes about 200 nm in diameter, and the quantity of loaded genes highly increased by more than 8.8-fold. The liposome membrane was composed of neutral lipids free from cationic charges. Consequently, this system is nontoxic, unlike other traditional cationic gene carriers. Intense red fluorescent protein (RFP) expression in CHO-K1 cells showed that the amplified genes could be successfully transfected to cells. Animal experiments with the luciferase gene also showed in vivo gene expression by our system without toxicity. We think that this PCR-based nanofactory system can overcome the toxicity problem that is the critical limitation of current gene delivery to clinical application.
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%
SAFC
Cholesterol, Plant-Derived, SyntheChol®, NF, Ph. Eur., JP
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications