  • Search Results
  • Efficient and sustained IGF-1 expression in the adipose tissue-derived stem cells mediated via a lentiviral vector.
Efficient and sustained IGF-1 expression in the adipose tissue-derived stem cells mediated via a lentiviral vector.
Journal of molecular histology (2014-12-04)
Ting Chen, Dangsheng Huang, Guanghui Chen, Tingshu Yang, Jun Yi, Miao Tian
ABSTRACT
The adipose tissue-derived stem cells (ADSCs) represent a significant area of the cell therapy. Genetic modification of ADSCs may further improve their therapeutic potential. Here, we aimed to generate a lentiviral vector expressing insulin-like growth factor-I (IGF-1) and investigate the impact of IGF-1 transduction on the properties of cultured ADSCs. Isolated rat ADSCs were assessed by flow cytometric analysis. IGF-1 was cloned and inserted into the pLenO-DCE plasmid to acquire pLenO-DCE-IGF-1 plasmid. Lentivirus was enveloped with pRsv-REV, pMDlg-pRRE and pMD2G plasmids in 293T cells. The ADSCs were transfected with the vectors. And then IGF-1-induced anti-apoptosis was evaluated by annexin V-FITC. Besides, proliferation of cells was detected by MTT assay and EdU. Moreover, Akt phosphorylation was evaluated by Western blotting analysis. Stable expression of IGF-1 in ADSCs was confirmed. ADSCs were positive for CD90 and CD29, but negative for CD31, CD34 and CD45. The transduction of IGF-1 to the ADSCs caused a dramatic increase in P-Akt expression. Over-expression of IGF-1 in ADSCs could improve the paracine of IGF-1 in a time-dependent manner, but could not promote the proliferation of ADSCs. This study indicated that lentiviral vectors offered a promising mean of delivering IGF-1 to the ADSCs. Lentiviral-mediated over-expression of therapeutic IGF-1 gene in ADSCs could prolong the anti-apoptosis effect of IGF-1, which might be induced by the activation of the PI3K/Akt pathway. And our data would improve the efficacy of ADSC-based therapies.
Product Description
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Calcitriol, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)