  • Search Results
  • Anti-inflammatory, anticholinesterase and antioxidant activity of leaf extracts of twelve plants used traditionally to alleviate pain and inflammation in South Africa.
Anti-inflammatory, anticholinesterase and antioxidant activity of leaf extracts of twelve plants used traditionally to alleviate pain and inflammation in South Africa.
Journal of ethnopharmacology (2014-12-06)
J P Dzoyem, J N Eloff
ABSTRACT
Oxidative stress and inflammatory conditions are among the pathological features associated with the central nervous system in Alzheimer׳s disease. Traditionally, medicinal plants have been used to alleviate inflammation, pains and also other symptoms possibly associated with Alzheimer׳s disease. Therefore, the present study was designed to determine the in vitro anti-inflammatory, antioxidant and anticholinesterase activity of twelve South African medicinal plants traditionally used to alleviate pain and inflammation. Nitric oxide (NO) production in LPS-activated RAW 264.7 macrophages and 15-lipoxygenase (LOX) inhibitory assay were used to evaluate the anti-inflammatory activity. Acetylcholinesterase inhibition was assessed by using a modification of the Ellman׳s method. Antioxidant activity, total phenolic and total flavonoids contents were determined using standard in vitro methods. The extract of Burkea africana had the highest anti-15-lipoxygenase activity with 85.92% inhibition at 100µg/mL. All the extracts tested inhibited nitric oxide (NO) production in a dose dependant manner in LPS-stimulated RAW 264.7 macrophages. However, extracts from Leucaena leucocephala, Lippia javanica inhibited the production of NO by 97% at a concentration of 25µg/mL. In addition, both Leucaena leucocephala and Englerophytum magaliesmontanum had strong activity against acetylcholinesterase with IC50 values of 118µg/mL and 160µg/mL respectively. Hight levels of phenolics and flavonoids were found in Leucaena leucocephala, Lippia javanica and Burkea africana. The correlation with antioxidant activities was not strong indicating that other metabolites may also be involved in antioxidant activity. The results obtained in this study validate the use of leaf extracts of these plants in South African traditional medicine against inflammation. Extracts of these plants species might be of value in the management of various diseases emerging from oxidative stress and related degenerative disorders.
Product Description
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
L-Glutamine
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Supelco
Gallic acid, certified reference material, TraceCERT®
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®
SAFC
Tromethamine
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Sigma-Aldrich
Potassium persulfate, ACS reagent, ≥99.0%
Trometamol, European Pharmacopoeia (EP) Reference Standard
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)