  • Search Results
  • Effect of power ultrasound pretreatment on peptidic profiles and angiotensin converting enzyme inhibition of milk protein concentrate hydrolysates.
Effect of power ultrasound pretreatment on peptidic profiles and angiotensin converting enzyme inhibition of milk protein concentrate hydrolysates.
Journal of the science of food and agriculture (2014-01-15)
Hankie Uluko, Lu Liu, Hongjuan Li, Wenming Cui, Shuwen Zhang, Lili Zhao, Haixiao Xue, Jiaping Lv
ABSTRACT
The use of power ultrasound as a pretreatment to enhance the hydrolysis of milk protein concentrate (MPC) and subsequent angiotensin converting enzyme (ACE) inhibitory activity has been studied. Liquid chromatography was used to analyse peptide profiles of Neutrase-derived MPC hydrolysates after pretreatment at 0, 1, 3, 5 and 8 min at an ultrasound power level of 800 W. The peptide profiles indicated an increase in number of peptides when ultrasound pretreatment was applied. There was also an increase in the degree of hydrolysis of MPC hydrolysates. The profiles indicated that new small peptides in ultrasound pretreated samples (1-5 min) which were not present in the control samples and 8 min pretreated samples, could be responsible for increased ACE inhibitory activity. These small peptides were digested in the 8 min pretreated samples. Ultrasound pretreatment of MPC increases the ACE inhibitory activity of the hydrolysates because of the production of new small peptides. This can be used as a means to derive potent ACE inhibitory peptides at industrial scale in complex protein sources.
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly
Serine, European Pharmacopoeia (EP) Reference Standard