MilliporeSigma
Inhibiting protein arginine deiminases has antioxidant consequences.
The Journal of pharmacology and experimental therapeutics (2015-01-31)
Erin E Witalison, Xiangli Cui, Anne B Hofseth, Venkataraman Subramanian, Corey P Causey, Paul R Thompson, Lorne J Hofseth
ABSTRACT
Ulcerative colitis is a dynamic, idiopathic, chronic inflammatory condition that carries a high colon cancer risk. We previously showed that Cl-amidine, a small-molecule inhibitor of the protein arginine deiminases, suppresses colitis in mice. Because colitis is defined as inflammation of the colon associated with infiltration of white blood cells that release free radicals and citrullination is an inflammation-dependent process, we asked whether Cl-amidine has antioxidant properties. Here we show that colitis induced with azoxymethane via intraperitoneal injection + 2% dextran sulfate sodium in the drinking water is suppressed by Cl-amidine (also given in the drinking water). Inducible nitric oxide synthase, an inflammatory marker, was also downregulated in macrophages by Cl-amidine. Because epithelial cell DNA damage associated with colitis is at least in part a result of an oxidative burst from overactive leukocytes, we tested the hypothesis that Cl-amidine can inhibit leukocyte activation, as well as subsequent target epithelial cell DNA damage in vitro and in vivo. Results are consistent with this hypothesis, and because DNA damage is a procancerous mechanism, our data predict that Cl-amidine will not only suppress colitis, but we hypothesize that it may prevent colon cancer associated with colitis.
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nitric oxide, 98.5%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy