MilliporeSigma
Comprehensive profiling and natural variation of flavonoids in rice.
Journal of integrative plant biology (2014-04-16)
Xuekui Dong, Wei Chen, Wensheng Wang, Hongyan Zhang, Xianqing Liu, Jie Luo
ABSTRACT
Flavonoids constitute a major group of plant phenolic compounds. While extensively studied in Arabidopsis, profiling and naturally occurring variation of these compounds in rice (Oryza sativa), the monocot model plant, are less reported. Using a collection of rice germplasm, comprehensive profiling and natural variation of flavonoids were presented in this report. Application of a widely targeted metabolomics method facilitated the simultaneous identification and quantification of more than 90 flavonoids using liquid chromatography tandem mass spectrometry (LC-MS/MS). Comparing flavonoid contents in various tissues during different developmental stages revealed tissue-specific accumulation of most flavonoids. Further investigation indicated that flavone mono-C-glycosides, malonylated flavonoid O-hexosides, and some flavonoid O-glycosides accumulated at significantly higher levels in indica than in japonica, while the opposite was observed for aromatic acylated flavone C-hexosyl-O-hexosides. In contrast to the highly differential accumulation between the two subspecies, relatively small variations within subspecies were detected for most flavonoids. Besides, an association analysis between flavonoid accumulation and its biosynthetic gene sequence polymorphisms disclosed that natural variation of flavonoids was probably caused by sequence polymorphisms in the coding region of flavonoid biosynthetic genes. Our work paves the way for future dissection of biosynthesis and regulation of flavonoid pathway in rice.
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid solution, suitable for HPLC