  • Search Results
  • Differential efficacy of human mesenchymal stem cells based on source of origin.
Differential efficacy of human mesenchymal stem cells based on source of origin.
Journal of immunology (Baltimore, Md. : 1950) (2014-10-03)
Erin Collins, Fei Gu, Maosong Qi, Ivan Molano, Phillip Ruiz, Lingyun Sun, Gary S Gilkeson
ABSTRACT
Mesenchymal stem cells (MSCs) are useful in tissue repair but also possess immunomodulatory properties. Murine and uncontrolled human trials suggest efficacy of MSCs in treating lupus. Autologous cells are preferable; however, recent studies suggest that lupus-derived MSCs lack efficacy in treating disease. Thus, the optimum derivation of MSCs for use in lupus is unknown. It is also unknown which in vitro assays of MSC function predict in vivo efficacy. The objectives for this study were to provide insight into the optimum source of MSCs and to identify in vitro assays that predict in vivo efficacy. We derived MSCs from four umbilical cords, four healthy bone marrows (BMs), and four lupus BMs. In diseased MRL/lpr mice, MSCs from healthy BM and umbilical cords significantly decreased renal disease, whereas lupus BM MSCs only delayed disease. Current in vitro assays did not differentiate efficacy of the different MSCs. However, differences in MSC efficacy were observed in B cell proliferation assays. Our results suggest that autologous MSCs from lupus patients are not effective in treating disease. Furthermore, standard in vitro assays for MSC licensing are not predictive of in vivo efficacy, whereas inhibiting B cell proliferation appears to differentiate effective MSCs from ineffective MSCs.
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)