  • Search Results
  • Surfactant proteins A, B, C and D in the human nasal airway: associated with mucosal glands and ciliated epithelium but absent in fluid-phase secretions and mucus.
Surfactant proteins A, B, C and D in the human nasal airway: associated with mucosal glands and ciliated epithelium but absent in fluid-phase secretions and mucus.
ORL; journal for oto-rhino-laryngology and its related specialties (2014-12-17)
Maria Q Gaunsbaek, Anette D Kjeldsen, Viggo Svane-Knudsen, Maiken L Henriksen, Soren Hansen
ABSTRACT
To investigate the presence of surfactant protein (SP) A, B, C and D in nasal airways and to determine whether the proteins exert their main functions in nasal secretions or in the deeper layers of the nasal mucosa. Volunteers were recruited from the Department of ENT Head and Neck Surgery, Odense University Hospital, Denmark. The study included 39 subjects. Nasal mucosal biopsies were analyzed by immunohistochemistry, and bronchoalveolar and nasal lavages, nasal brush biopsies and nasal mucus were analyzed for SP-A, -B, -C and -D by SDS-PAGE and Western blotting. The presence of SP-A and SP-D in the first three samplings were also analyzed by enzyme-linked immunosorbent assay. In nasal mucosal biopsies, SP-A, -B, -C and -D were all demonstrated in the serous acini of the submucosal glands and in the surface epithelium. SP-D was detected in nasal brush biopsies, whereas the other SPs were absent. Moreover, SP-A, -B, -C and -D were absent in nasal lavage and mucus. SP-A, -B, -C and -D exert their protective effect in the ductal epithelium of the submucosal glands rather than in nasal secretions and mucus. Further studies are required to clarify the functions of these proteins in nasal secretory pathways for understanding upper airway diseases.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Eosin Y
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Eosin Y disodium salt, Dye content ≥85 %
Sigma-Aldrich
Eosin Y disodium salt, certified by the Biological Stain Commission
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O