  • Search Results
  • Bicarbonate enhances the in vitro antibiotic activity of kanamycin in Escherichia coli.
Bicarbonate enhances the in vitro antibiotic activity of kanamycin in Escherichia coli.
Letters in applied microbiology (2015-01-15)
M Gutiérrez-Huante, H Martínez, V H Bustamante, J L Puente, J Sánchez
ABSTRACT
Growth of enteropathogenic Escherichia coli E2348/69 was inhibited by bicarbonate in a dose-dependent manner, showing approximately 5% growth reduction at 5 mmol l(-1) while kanamycin at 3·12 μg ml(-1) inhibited growth by 15%, yet when kanamycin and bicarbonate were combined at these concentrations, inhibition increased to 80%. Unexpectedly, at bicarbonate concentrations >20 mmol l(-1) enhancement of the antibiotic activity virtually disappeared, i.e. there was a paradoxical Eagle-like effect. How bicarbonate acts is unclear, but neutral or alkaline pH also enhanced the activity of kanamycin. However, several differences indicated a separate effect of bicarbonate. First, bicarbonate inhibited growth more than the corresponding increments in pH. Second, at low concentration, the antibiotic enhancing effect of bicarbonate was stronger than the effect of pH alone. Third, 5 mmol l(-1) bicarbonate significantly enhanced the activity of kanamycin while the corresponding pH had no effect. Fourth, the Eagle-like effect was exclusive of bicarbonate because changes in pH did not induce an analogous behaviour. Notwithstanding the mechanism, the enhancing effect of bicarbonate was indubitable. Consequently, it seems worthwhile to explore further its potential to improve the efficacy of aminoglycosides and maybe even other antibiotics. Bicarbonate at a low concentration enhanced the in vitro antibiotic activity of kanamycin and gentamicin. Even though the action mechanism of bicarbonate is hitherto unknown, it seems worthwhile to explore further its capacity to improve the efficacy of aminoglycosides. Clearly, the well-known harmful side-effects of aminoglycosides are a concern. However, it has recently been shown in a fish model that bicarbonate may protect ciliary cells against the damage caused by aminoglycosides. So, it seems possible that bicarbonate could help reduce aminoglycoside dosage at the same time that it might help lessen the damage to auditory ciliary cells in humans.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, powder, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Kanamycin sulfate Streptomyces kanamyceticus, powder, γ-irradiated
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, meets USP testing specifications, powder
Sigma-Aldrich
Kanamycin sulfate from Streptomyces kanamyceticus, Animal Component-free