  • Search Results
  • mTOR limits the recruitment of CD11b+Gr1+Ly6Chigh myeloid-derived suppressor cells in protecting against murine immunological hepatic injury.
mTOR limits the recruitment of CD11b+Gr1+Ly6Chigh myeloid-derived suppressor cells in protecting against murine immunological hepatic injury.
Journal of leukocyte biology (2014-02-27)
Yan Zhang, Yujing Bi, Hui Yang, Xi Chen, Huanrong Liu, Yun Lu, Zhengguo Zhang, Jiongbo Liao, Shan Yang, Yiwei Chu, Ruifu Yang, Guangwei Liu
ABSTRACT
The mTOR pathway integrates diverse environmental inputs, including immune signals and metabolic cues, to direct the innate and adaptive immune responses. MDSCs are a heterogeneous cell population that plays a crucial regulatory effect in immune-related diseases. However, whether mTOR signaling affects the functions of MDSCs remains largely unknown. Here, we show that mTOR signaling is a pivotal negative determinant of MDSC recruitment in IMH disease. In the context of IMH, inhibition of mTOR with rapamycin in CD11b⁺Gr1⁺ MDSCs mediates protection against IMH and serves as a functional, suppressive immune modulator that results in increased CD11b⁺Gr1⁺Ly6C(high) MDSC recruitment to inflammatory sites. In agreement with this, mTOR down-regulation promotes CD11b⁺Gr1⁺Ly6C(high) MDSC migration in vitro and in vivo. Mechanistically, mTOR activity down-regulation in MDSCs induced iNOS expression and NO production. Pharmacologic inhibition of iNOS completely eliminated MDSC recruitment. This study identifies MDSCs as an essential component for protection against IMH following rapamycin treatment. Rapamycin treatment or mTOR inhibition promotes CD11b⁺Gr1⁺Ly6C(high) MDSC recruitment and is critically required for protection against hepatic injury. This study further validates the targeting of mTOR signaling as a potential therapeutic approach to IMH-related diseases.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1