Mesenchymal stem cells mitigate cirrhosis through BMP7.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-01-24)
Bing Li, Qing Shao, Dong Ji, Fan Li, Guofeng Chen
ABSTRACT
Transplantation of mesenchymal stem cells (MSCs) has therapeutic effects on various diseases, while its effect on developing cirrhosis as well as the underlying mechanism remained largely unknown. Twenty C57BL/6 mice were randomly separated into 2 groups of ten each. One group received transplantation of MSCs, while the other group received saline as control. The mice then received intraperitoneal injection of carbon tetrachloride (CCl4) twice per week for 8 weeks to develop cirrhosis. After another 4 weeks, the levels of cirrhosis in these mice were evaluated by liver fibrosis area, portal pressure, sodium balance and excretion. Transcripts of transforming growth factor β 1 (TGFβ1) and bone morphogenic protein 7 (BMP7) in the mouse livers were quantified by RT-qPCR. BMP7-depleted MSCs were prepared and applied in this model, and compared to MSCs. Liver fibrosis, portal hypertension and sodium retention that were developed by CCl4, were all significantly alleviated by MSCs transplantation, which decreased TGFβ1 levels and increased BMP7 levels in the injured liver. MSCs were found to express extremely high levels of BMP7. Knockdown of BMP7 in MSCs completely abolished the protective effect of MSCs against CCl4-induced cirrhosis. MSCs mitigate cirrhosis through their production of BMP7 against the fibrogenic effect of TGFβ1 in the injured liver.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Carbon tetrachloride, reagent grade, 99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Carbon tetrachloride, suitable for HPLC, ≥99.9%
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Cholecalciferol (D3), analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Supelco
Carbon tetrachloride, analytical standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, analytical standard