MilliporeSigma
Soluble ST2 reflects hemodynamic stress in non-ischemic heart failure.
International journal of cardiology (2014-12-03)
Kaspar Broch, Arne K Andreassen, Thor Ueland, Annika E Michelsen, Wenche Stueflotten, Pål Aukrust, Svend Aakhus, Lars Gullestad
ABSTRACT
Elevated levels of soluble ST2 (sST2) are associated with adverse outcome in heart failure. A change in sST2 levels has also been shown to presage outcome. In vitro, ST2 expression is induced by myocardial stress and pro-inflammatory stimuli. The determinants of sST2 levels in vivo, and how they vary with clinical status over time, have not been well described. In a cohort of patients with non-ischemic heart failure, we aimed to assess the association between sST2-levels and hemodynamic parameters reflecting right and left ventricular pre- and afterload, and how these vary with time and clinical status. We prospectively recruited 102 patients with a left ventricular ejection fraction of 26 ± 10% and a diagnosis of idiopathic dilated cardiomyopathy based on patient history, clinical examination, echocardiography and coronary angiography. Patients went through extensive baseline work-up and were re-examined after one year. Subsequently, heart transplantations and deaths were recorded. Determinants of sST2 were analyzed at baseline and after one year. Soluble ST2 was measured with a highly sensitive immunoassay. Soluble ST2 levels were associated with hemodynamic parameters, but these associations were attenuated with clinical improvement. Soluble ST2 was elevated in patients with severe symptoms, but did not vary with etiology, viral presence or the amount of myocardial fibrosis. Heart rate and right atrial pressure remained independent predictors of sST2 on multiple regression analysis. Our results imply that in non-ischemic heart failure, sST2 reflects hemodynamic stress rather than pathogenic processes in the myocardium.
Product Description
Supelco
Meglumine, Pharmaceutical Secondary Standard; Certified Reference Material
Meglumine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Meglumine, meets USP testing specifications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
N-Methyl-D-glucamine, 99.0-100.5% (titration)
Sigma-Aldrich
N-Methyl-D-glucamine, ReagentPlus®, ≥99.0% (T)