  • Search Results
  • Oncogenic roles of Bmi1 and its therapeutic inhibition by histone deacetylase inhibitor in tongue cancer.
Oncogenic roles of Bmi1 and its therapeutic inhibition by histone deacetylase inhibitor in tongue cancer.
Laboratory investigation; a journal of technical methods and pathology (2014-10-07)
Zhongwu Li, Yanling Wang, Chunping Yuan, Yumin Zhu, Jing Qiu, Wei Zhang, Bing Qi, Heming Wu, Jinhai Ye, Hongbing Jiang, Jianrong Yang, Jie Cheng
ABSTRACT
The polycomb complex protein Bmi1 (B lymphoma Mo-MLV insertion region 1 homolog) mediates epigenetic transcriptional silencing by modifying chromatin structure and is critical for stem cell homeostasis and tumorigenesis. Bmi1 is frequently overexpressed in human malignancies and therefore has key diagnostic and prognostic significance, and holds potential as a therapeutic target. Here we sought to characterize the expression patterns and oncogenic roles of Bmi1 in tongue squamous cell carcinoma and to determine the anticancer effects of histone deacetylase inhibitors (HDACis) via Bmi1 inhibition against tongue cancer. Our data revealed that Bmi1 was aberrantly overexpressed in a significant portion of tongue cancers. Elevated Bmi1 is associated with cervical node metastasis, Ki-67 abundance and reduced overall survival, and also serves as an independent prognostic factor for patient outcomes. Short-hairpin RNA-mediated Bmi1 knockdown inhibited cell proliferation and migration, induced cell apoptosis and senescence, reduced colony formation and CD44(+)CD133(+) sub-population as well as enhanced cisplatin chemosensitivity, presumably by modulation of p16, p14 and E-cadherin. Moreover, HDACi chemicals Trichostatin A (TSA) and sodium butyrate (NaB) potently inhibited Bmi1 and triggered similar phenotypic changes reminiscent of Bmi1 silencing, although TSA treatment seemed paradoxically to induce some epithelial-mesenchymal transition-like changes in tongue cancer cells. Importantly, NaB-induced antitumor effects were partially attenuated by enforced Bmi1 overexpression in vitro. Genetic Bmi1 silencing and pharmacological inhibition of Bmi1 by NaB treatment significantly impaired tumor growth in a tongue cancer xenograft model. Taken together, our results indicate that Bmi1 serves as a key driver and biomarker with multiple oncogenic functions underlying tongue tumorigenesis. Selected appropriate HDACi compounds like NaB may represent novel therapeutic agents against tongue cancer.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O