  • Search Results
  • The influence of ezetimibe on classical and alternative activation pathways of monocytes/macrophages isolated from patients with hypercholesterolemia.
The influence of ezetimibe on classical and alternative activation pathways of monocytes/macrophages isolated from patients with hypercholesterolemia.
Naunyn-Schmiedeberg's archives of pharmacology (2014-05-02)
Dariusz Suchy, Krzysztof Łabuzek, Grzegorz Machnik, Bogusław Okopień
ABSTRACT
Macrophages are crucial for the development of atherosclerotic plaques. Classically activated macrophages contribute to plaque growth and destabilization, while alternatively activated macrophages increase plaque stability. Here, we assessed the influence of ezetimibe on the activation of monocyte-derived macrophages isolated from patients with hypercholesterolemia (total cholesterol 263.4 ± 12.5 mg/dl, low-density lipoprotein cholesterol 179.7 ± 11.3 mg/dl, triglycerides 123.9 ± 11.4 mg/dl). Cells were stimulated with 1 μg/ml lipopolysaccharide (LPS) or 1 μg/ml LPS plus 22 ng/ml ezetimibe. Control cells were left unstimulated. The expression of classical activation markers (interleukin-1β (IL-1β), nitric oxide (NO), and inducible nitric oxide synthase (iNOS)) and alternative activation markers (mannose receptor (MR) and arginase-1 (Arg1)) was determined after 48 h. The employed analytical methods included enzyme-linked immunosorbent assay, Griess reaction, real-time polymerase chain reaction, and Western blotting. LPS increased the secretion of IL-1β and NO and the expression of iNOS mRNA, iNOS protein, and Arg1 protein. It did not affect the expression of MR or Arg1 mRNA. In comparison to LPS stimulation, co-stimulation with ezetimibe decreased the secretion of IL-1β and the expression of iNOS mRNA and protein, while it increased MR mRNA and protein expression. Co-stimulation with ezetimibe did not change the secretion of NO or the expression of Arg1. The results suggest that ezetimibe in inflammatory in vitro conditions contributes to the suppression of classical and promotion of the alternative macrophage activation.
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture