  • Search Results
  • A human dendritic cell-based in vitro model to assess Mycobacterium tuberculosis SO2 vaccine immunogenicity.
A human dendritic cell-based in vitro model to assess Mycobacterium tuberculosis SO2 vaccine immunogenicity.
ALTEX (2014-05-21)
Marilena P Etna, Elena Giacomini, Martina Severa, Manuela Pardini, Nacho Aguilo, Carlos Martin, Eliana M Coccia
ABSTRACT
Among the tuberculosis (TB) vaccine candidates, SO2 is the prototype of the first live-attenuated vaccine that recently entered into clinical trials. To investigate the capacity of SO2 to stimulate an appropriate immune response in vitro within a human immunological context, a comparative analysis of the effects promoted by SO2, the current Bacille Calmette-Guerin (BCG) vaccine and Mycobacterium tuberculosis (Mtb) was conducted in human primary dendritic cells (DC), which are critical modulators of vaccine-induced immunity. In particular, we found that SO2 promotes the expression of maturation markers similarly to BCG but at a lower extent than Mtb. Moreover, SO2-infected DC released higher levels of interleukin (IL)-23 than BCG-infected cells, which account for the expansion of interferon (IFN)-γ-producing T cells in an IL-12-independent manner. In the autologous mixed leukocyte reaction setting, the expansion of IL-17-producing T cells was also observed in response to SO2 infection. Interestingly, apoptosis and autophagic flux, events required for the antigen presentation within MHC class II complex, were not affected in DC infected with SO2, conversely to what observed upon Mtb stimulation. Collectively, our results indicate that SO2 represents a promising TB vaccine candidate, which displays an attenuated phenotype and promotes in DC a stronger capacity to stimulate the Th response than BCG vaccine. Interestingly, the data obtained by using the human DC-based experimental setting mirrored the results derived from studies in animal models, suggesting that this system could be used for an efficient and rapid down-selection of new TB vaccine candidates, contributing to achieve the "3Rs" objective.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems
SAFC
BIS-TRIS
SAFC
BIS-TRIS
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O