  • Search Results
  • The mammalian target of rapamycin complex 1 (mTORC1) in breast cancer: the impact of oestrogen receptor and HER2 pathways.
The mammalian target of rapamycin complex 1 (mTORC1) in breast cancer: the impact of oestrogen receptor and HER2 pathways.
Breast cancer research and treatment (2015-02-24)
Dena A Jerjees, Ola H Negm, M Layth Alabdullah, Sameer Mirza, Methaq Alkaabi, Mohamed R Hameed, Rezvan Abduljabbar, Abir Muftah, Chris C Nolan, Andrew R Green, Patrick J Tighe, Vimla Band, Ian O Ellis, Emad A Rakha
ABSTRACT
The mammalian target of rapamycin complex 1 (mTORC1) is a downstream of the PI3K/Akt pathway which affects cancer development. mTORC1 has many downstream signalling effectors that can enhance different cellular responses. This study aims to investigate the expression of mTORC1 in breast cancer (BC) and correlate it with key clinicopathological and molecular features of BC especially to proteins related to oestrogen receptor (ER) and HER2 pathways in different BC classes. Moreover, mTORC1 expression was assessed in 6 BC cell lines including ER+ and ER- cell lines with and without HER2 transfection. Immunohistochemistry was used to assess the expression of phospho (p) mTORC1 in a large (n = 1300) annotated BC series prepared as tissue microarray. Reverse phase protein array (RPPA) was used to assess its expression in the different BC cell lines. The expression of p-mTORC1 was cytoplasmic with moderate/high expression noted in 44 % of BC. p-mTORC1 expression was associated with clinicopathological variables characteristic of good prognosis. Positive correlation with ER, ER-related proteins AKT, PI3K and luminal differentiation markers were observed in the whole series and in the ER+HER2- subgroup. Association with HER2 was mainly observed in the ER-negative class. RPPA indicated that p-mTORC1 expression was mainly related to ER expression and with better outcome in the Akt positive tumours. p-mTORC1 is associated with good prognostic features. Its expression is related to ER and ER related proteins in addition to AKT and PI3K. Its relation with HER2 expression is mainly seen in the absence of ER expression.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
o-Xylene, puriss. p.a., ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
o-Xylene, analytical standard
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Social Media
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
MilliporeSigma
Research. Development. Production.
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.
Reproduction of any materials from the site is strictly forbidden without permission.