  • Search Results
  • Selective anticancer effects and protection from chemotherapy by the botanical compound LCS101: Implications for cancer treatment.
Selective anticancer effects and protection from chemotherapy by the botanical compound LCS101: Implications for cancer treatment.
International journal of oncology (2014-10-22)
Zoya Cohen, Yair Maimon, Merav Yoeli-Lerner, Peiying Yang, Noah Samuels, Raanan Berger
ABSTRACT
There is a need for new options for reducing the side effects of cancer treatment, without compromising efficacy, enabling patients to complete treatment regimens. The botanical compound LCS101 exhibits inhibitory effects on cancer cell growth, and reduces chemotherapy-induced hematological toxicities. The aim of the present study is to examine the selectivity of the effects of the compound, alone and in conjunction with conventional chemotherapy agents, on cancer cell proliferation. The effects of LCS101 were tested on a number of cancer cell lines (breast, MCF7, MDA-MB‑231; colorectal, HCT116; prostate, PC-3, DU-145) and on non-tumorigenic normal human epithelial cells (breast, MCF10A; prostate, EP#2). Cell viability was analyzed using an XTT assay and observed by light microscopy. Necrosis and apoptosis were examined using FACS analysis and immunoblotting. LCS101 selectively induced cell death in breast, colon and prostate cancer cell lines, as measured by XTT assay. Light microscopy and FACS analysis showed changes indicative of a necrotic process. LCS101 was also found to induce PARP-1 reduction in breast cancer cells, with no effect on non-tumorigenic breast epithelial cells. While LCS101 increased cell death in cancer cells exposed to doxorubicin and 5-FU, it showed a protective effect on non-tumorigenic human epithelial cells from chemotherapy-induced cell death. A similar selective effect was observed with apoptosis-associated PARP-1 cleavage. The findings demonstrate that the anti-proliferative effects exhibited by the botanical compound LCS101 are selective to cancer cells, and offer protection to non-tumorigenic normal epithelial cells from chemotherapy agents.
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Deoxycholic acid, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)