  • Search Results
  • Promoting thiol expression increases the durability of antitumor T-cell functions.
Promoting thiol expression increases the durability of antitumor T-cell functions.
Cancer research (2014-08-29)
Pravin Kesarwani, Amir A Al-Khami, Gina Scurti, Krishnamurthy Thyagarajan, Navtej Kaur, Shahid Husain, Quan Fang, Osama S Naga, Patricia Simms, Gyda Beeson, Christina Voelkel-Johnson, Elizabeth Garrett-Mayer, Craig C Beeson, Michael I Nishimura, Shikhar Mehrotra
ABSTRACT
Ex vivo-expanded CD8(+) T cells used for adoptive immunotherapy generally acquire an effector memory-like phenotype (TEM cells). With regard to therapeutic applications, two undesired features of this phenotype in vivo are limited persistence and reduced antitumor efficacy, relative to CD8(+) T cells with a central memory-like phenotype (TCM cells). Furthermore, there is incomplete knowledge about all the differences between TEM and TCM cells that may influence tumor treatment outcomes. Given that TCM cells survive relatively longer in oxidative tumor microenvironments, we investigated the hypothesis that TCM cells possess relatively greater antioxidative capacity than TEM cells. Here, we report that TCM cells exhibit a relative increase compared with TEM cells in the expression of cell surface thiols, a key target of cellular redox controls, along with other antioxidant molecules. Increased expression of redox regulators in TCM cells inversely correlated with the generation of reactive oxygen and nitrogen species, proliferative capacity, and glycolytic enzyme levels. Notably, T-cell receptor-transduced T cells pretreated with thiol donors, such as N-acetyl cysteine or rapamycin, upregulated thiol levels and antioxidant genes. A comparison of antitumor CD8(+) T-cell populations on the basis of surface thiol expression showed that thiol-high cells persisted longer in vivo and exerted superior tumor control. Our results suggest that higher levels of reduced cell surface thiols are a key characteristic of T cells that can control tumor growth and that profiling this biomarker may have benefits to adoptive T-cell immunotherapy protocols.
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Interleukin-15 human, >97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
IL-15 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
IL-15 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
IL-15 from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)