  • Search Results
  • Effect-directed analysis and mixture effects of AhR-active PAHs in crude oil and coastal sediments contaminated by the Hebei Spirit oil spill.
Effect-directed analysis and mixture effects of AhR-active PAHs in crude oil and coastal sediments contaminated by the Hebei Spirit oil spill.
Environmental pollution (Barking, Essex : 1987) (2015-02-04)
Seongjin Hong, Sangwoo Lee, Kyungho Choi, Gi Beum Kim, Sung Yong Ha, Bong-Oh Kwon, Jongseong Ryu, Un Hyuk Yim, Won Joon Shim, Jinho Jung, John P Giesy, Jong Seong Khim
ABSTRACT
The major AhR-active PAHs were identified in crude oil and oil-contaminated sediments by use of effect-directed analysis. As part of the study, an enhanced potency balance analysis was conducted by establishing the novel relative potency values of (alkyl)-PAHs from the H4IIE-luc bioassay. Silica gel column fractionation of crude oil resulted in greater AhR-mediated potencies in fractions of aromatics (F2) and resins (F3), and such trend was also observed for field collected sediment samples. AhR-mediated potencies of six F2 sub-fractions from HPLC indicated that the majority of F2 responses were attributable to 3-4 ring aromatics. Target PAHs including C4-phenanthrene, C1-chrysene, and C3-chrysene in sediments explained ∼ 18% of the bioassay-derived TCDD-EQs, however, the unknown AhR agonists and potential mixture effects remain in question. Overall, the AhR-potency and antagonistic potential of residual oil in sediment tended to decrease over time, thus monitoring of weathering process would be key for the post management of oil-contaminated sites.
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Dichloromethane, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)