  • Search Results
  • Distinct mechanisms of cell-kill by triapine and its terminally dimethylated derivative Dp44mT due to a loss or gain of activity of their copper(II) complexes.
Distinct mechanisms of cell-kill by triapine and its terminally dimethylated derivative Dp44mT due to a loss or gain of activity of their copper(II) complexes.
Biochemical pharmacology (2014-08-19)
Kimiko Ishiguro, Z Ping Lin, Philip G Penketh, Krishnamurthy Shyam, Rui Zhu, Raymond P Baumann, Yong-Lian Zhu, Alan C Sartorelli, Thomas J Rutherford, Elena S Ratner
ABSTRACT
Triapine, currently being evaluated as an antitumor agent in phase II clinical trials, and its terminally dimethylated derivative Dp44mT share the α-pyridyl thiosemicarbazone backbone that functions as ligands for transition metal ions. Yet, Dp44mT is approximately 100-fold more potent than triapine in cytotoxicity assays. The aims of this study were to elucidate the mechanisms underlying their potency disparity and to determine their kinetics of cell-kill in culture to aid in the formulation of their clinical dosing schedules. The addition of Cu(2+) inactivated triapine in a 1:1 stoichiometric fashion, while it potentiated the cytotoxicity of Dp44mT. Clonogenic assays after finite-time drug-exposure revealed that triapine produced cell-kill in two phases, one completed within 20 min that caused limited cell-kill, and the other occurring after 16 h of exposure that produced extensive cell-kill. The ribonucleotide reductase inhibitor triapine at 0.4 μM caused immediate complete arrest of DNA synthesis, whereas Dp44mT at this concentration did not appreciably inhibit DNA synthesis. The inhibition of DNA synthesis by triapine was reversible upon its removal from the medium. Cell death after 16 h exposure to triapine paralleled the appearance of phospho-(γ)H2AX, a marker of DNA double-strand breaks induced by collapse of DNA replication forks after prolonged replication arrest. In contrast to triapine, Dp44mT produced robust cell-kill within 1h in a concentration-dependent manner. The short-term action of both agents was prevented by thiols, indicative of the involvement of reactive oxygen species. The time dependency in the production of cell-kill by triapine should be considered in treatment regimens.
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Hydroxycarbamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dp44mT, ≥98% (HPLC)
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)