  • Search Results
  • Rapid measurement of tacrolimus in whole blood by paper spray-tandem mass spectrometry (PS-MS/MS).
Rapid measurement of tacrolimus in whole blood by paper spray-tandem mass spectrometry (PS-MS/MS).
Clinica chimica acta; international journal of clinical chemistry (2014-12-30)
Run-Zhang Shi, El Taher M El Gierari, Nicholas E Manicke, James D Faix
ABSTRACT
Liquid chromatography-tandem mass spectrometry (LC-MS/MS) provides sensitivity and specificity for monitoring tacrolimus drug level in blood, but it requires an LC system and sample preparation, which is not amenable to random access testing typical of immunoassays. Paper spray (PS) ionization generates gas phase analyte ions directly from dried blood spots without sample preparation and LC. We evaluated a PS-MS/MS method for tacrolimus drug monitoring in a clinical diagnostic laboratory. Whole blood sample was mixed with stable isotope labeled internal standard ([(13)C, (2)H2]-FK506) and spotted onto a cartridge containing triangular shaped card paper. After drying, samples were analyzed by PS MS/MS in the selected reaction monitoring (SRM) mode, with a run time of 3 min/sample. Analytical measurement range was 1.5-30 ng/ml. Assay inter-day imprecision was 13%, 8%, and 5% at tacrolimus concentrations of 4.5, 10.5, and 24.5 ng/ml, respectively. Accuracy was determined by pure tacrolimus solution and was confirmed by result correlation to an immunoassay (slope=1.0, intercept=-0.02; r(2)=0.99), and to a conventional LC-MS/MS method (slope=0.90, intercept=0.4; r(2)=0.94). PS-MS/MS provides accurate results for tacrolimus with rapid turnaround time amenable to random access testing protocols.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%