  • Search Results
  • Discovery of MK-8970: an acetal carbonate prodrug of raltegravir with enhanced colonic absorption.
Discovery of MK-8970: an acetal carbonate prodrug of raltegravir with enhanced colonic absorption.
ChemMedChem (2014-12-04)
Abbas M Walji, Rosa I Sanchez, Sophie-Dorothee Clas, Rebecca Nofsinger, Manuel de Lera Ruiz, Jing Li, Amrithraj Bennet, Christopher John, David Jonathan Bennett, John M Sanders, Christina N Di Marco, Somang Hope Kim, Jaume Balsells, Scott S Ceglia, Qun Dang, Kimberly Manser, Becky Nissley, John S Wai, Michael Hafey, Junying Wang, Gene Chessen, Allen Templeton, John Higgins, Ronald Smith, Yunhui Wu, Jay Grobler, Paul J Coleman
ABSTRACT
Developing new antiretroviral therapies for HIV-1 infection with potential for less frequent dosing represents an important goal within drug discovery. Herein, we present the discovery of ethyl (1-((4-((4-fluorobenzyl)carbamoyl)-1-methyl-2-(2-(5-methyl- 1,3,4-oxadiazole-2-carboxamido)propan-2-yl)-6-oxo-1,6-dihydropyrimidin-5-yl)oxy)ethyl) carbonate (MK-8970), a highly optimized prodrug of raltegravir (Isentress). Raltegravir is a small molecule HIV integrase strand-transfer inhibitor approved for the treatment of HIV infection with twice-daily administration. Two classes of prodrugs were designed to have enhanced colonic absorption, and derivatives were evaluated in pharmacokinetic studies, both in vitro and in vivo in different species, ultimately leading to the identification of MK-8970 as a suitable candidate for development as an HIV therapeutic with the potential to require less frequent administration while maintaining the favorable efficacy, tolerability, and minimal drug-drug interaction profile of raltegravir.
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Supelco
Tetramethylsilane, analytical standard, for NMR spectroscopy, ACS reagent
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
Sigma-Aldrich
Water, suitable for HPLC