  • Search Results
  • Effects of 6-(4-chlorophenoxy)-tetrazolo[5,1-a]phthalazine on anticoagulation in mice and the inhibition of experimental thrombosis in rats.
Effects of 6-(4-chlorophenoxy)-tetrazolo[5,1-a]phthalazine on anticoagulation in mice and the inhibition of experimental thrombosis in rats.
Journal of cardiovascular pharmacology (2014-08-29)
Hai-Ling Yu, Feng Zhang, Tian Lan, Zhe-Shan Quan
ABSTRACT
: Thrombosis is a major complication that could be fatal in acute or chronic cardio-cerebral-vascular diseases. Therefore, the development of novel agents for anticlotting and the prevention of thrombosis and cardiovascular diseases are clinically significant. This study aimed to evaluate the anticoagulant and antithrombotic effects of 6-(4-chlorophenoxy)-tetrazolo[5,1-a]phthalazine (Q808), a new phthalazine tetrazole derivative. Bleeding time, clotting time, and serum calcium ion (Ca) concentration were assessed in mice, whereas arteriovenous thrombus weight and plasma prothrombin time were evaluated in rats, and platelets Ca influx was determined in rabbit. Daily oral administration of Q808 at 25, 50, or 100 mg/kg for 3 days significantly delayed bleeding time and clotting time in mice compared with controls. Q808 administration at 50 mg/kg significantly reduced experimental thrombus weight by 62.6% and delayed plasma prothrombin time by 58.7% in rats, whereas 50 and 100 mg/kg of Q808 daily significantly increased serum Ca concentration in mice. Q808 at 0.2, 0.4, and 0.8 mg/mL significantly inhibited thrombin-induced Ca influx in rabbit platelets. Our results suggest that Q808 at 25-200 mg/kg daily exerts anticoagulant and antithrombotic effects, and its mechanisms of action may involve both the intrinsic and extrinsic coagulation pathways that inhibit certain coagulation factors and platelet functions.
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%