  • Search Results
  • Preparation by coaxial electrospinning and characterization of membranes releasing (-) epicatechin as scaffold for tissue engineering.
Preparation by coaxial electrospinning and characterization of membranes releasing (-) epicatechin as scaffold for tissue engineering.
Materials science & engineering. C, Materials for biological applications (2014-12-11)
M M Castillo-Ortega, A G Montaño-Figueroa, D E Rodríguez-Félix, G Prado-Villegas, K P Pino-Ocaño, M J Valencia-Córdova, J M Quiroz-Castillo, P J Herrera-Franco
ABSTRACT
Optimal conditions for the preparation of a composite material of fibers of cellulose acetate (CA) and poly(vinyl pyrrolidone) (PVP), containing epicatechin (Epic) within the fiber CA/PVP-Epic/CA, were found. The morphology and physical/chemical properties of the fibrous membranes containing CA, PVP, and epicatechin were characterized using FTIR spectroscopy, thermal analysis, SEM, TEM, and natural weathering. Also, mechanical characterization of the fibers showed that tensile strength of the membrane was not affected by the presence of epicatechin within the fiber as compared with fibers without epicatechin. The effect of the medium on the release rate of epicatechin was also studied. The amount of epicatechin release was higher in water, 79.6%, and 31% in MesenCult medium. These results showed that these composite materials are recommended for cardiac tissue engineering; furthermore, using these materials allows precise release of the epicatechin in the damaged tissue.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Epicatechin, primary reference standard
Supelco
(−)-Epicatechin, analytical standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%