  • Search Results
  • Synthesis of variants of Marfey's reagent having d-amino acids as chiral auxiliaries and liquid-chromatographic enantioseparation of (RS)-Mexiletine in spiked plasma: assessment and comparison with L-amino acid analogs.
Synthesis of variants of Marfey's reagent having d-amino acids as chiral auxiliaries and liquid-chromatographic enantioseparation of (RS)-Mexiletine in spiked plasma: assessment and comparison with L-amino acid analogs.
Journal of chromatography. A (2015-01-13)
Ravi Bhushan, Vinod Kumar Vashistha
ABSTRACT
Five d-amino acids have been used for the first time to synthesize chiral derivatizing reagents (as variants of Marfey's reagent) by nucleophilic displacement of one of the fluorine atoms in 1,5-difluoro-2,4-dinitrobenzene as against the literature reports on application of only l-amino acids or their amides as chiral auxiliaries in dinitrobenzene (DNB) moiety. Five other DNB based reagents were also prepared by nucleophilic substitution of fluorine atom with the set of the same amino acids in l-configuration, as chiral auxiliaries. These reagents were characterized and used for synthesis of diastereomers of (RS)-Mexiletine spiked in human plasma. Diastereomers were synthesized employing microwave irradiation and were separated on reversed-phase C18 column. Performance of the two types of chiral derivatizing reagents was compared. The reagents containing d-amino acids provided enhanced separation of diastereomers than those containing l-amino acids. The best resolution was obtained using mobile phase consisting of acetonitrile and 0.1% trifluoroacetic acid in gradient mode. The detection was carried out at 340nm. The method so developed was validated for linearity, accuracy and precision. The limit of quantitation was found to be approximately 25.2ngmL(-1) in human plasma.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Supelco
Methanol, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, Absolute - Acetone free