  • Search Results
  • Selective expression of tumor necrosis factor-related apoptosis-inducing ligand mediated by microRNA suppresses renal carcinoma growth.
Selective expression of tumor necrosis factor-related apoptosis-inducing ligand mediated by microRNA suppresses renal carcinoma growth.
Molecular and cellular biochemistry (2014-05-03)
Zhuo Zhang, Haiyan Zhang, Hongyan Li, Xiaoliang Chen, Meihan Liu, Dayu Liu, Yuanyuan Zhao, Xiangbo Kong
ABSTRACT
Renal cell carcinoma (RCC) is the most common types among kidney cancers. Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) strongly induces apoptosis in RCC. However, TRAIL therapy also leads to hepatotoxicity. To improve the biosafety, we inserted miRNA response elements (MREs) of miR-138, miR-199, and miR-122 into an adenoviral vector, Ad-TRAIL-3MREs, to restrict TRAIL expression within RCC cells. Luciferase assays showed that MREs can regulate the expression of exogenous gene in RCC cells. Ad-TRAIL-3MREs selectively expressed TRAIL and induce apoptosis in RCC cells, but not in normal cells. MTT assays revealed that Ad-TRAIL-3MREs reduced viability of RCC cells without cytotoxicity to normal cells. Ad-TRAIL-3MREs suppressed the growth of ACHN tumors and exerted no hepatotoxicity in vivo. Collectively, we generated a TRAIL-expressing adenoviral vector under the regulation of MREs. This miRNA-based gene therapy may be a promising strategy for RCC treatment.
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)